Learn More
Invitrogen™ VANGL2 Polyclonal Antibody

Description
Sequence homology: Chimpanzee (100%), Opossum/Equine (93%), Chicken/Xenopus (73%).
Specifications
Specifications
| Antigen | VANGL2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | Vangl2 |
| Gene Accession No. | P84889, Q91ZD4, Q9ULK5 |
| Gene Alias | C530001F03Rik; KIAA1215; loop tail associated protein; loop-tail; Loop-tail protein 1; loop-tail protein 1 homolog; Loop-tail-associated protein; Lp; LPP1; Ltap; MGC119403; MGC119404; RGD:1309442}; ska>m17Jus>; ska17; STB1; stbm; STBM1; strabismus; Strabismus 1; van Gogh-like protein 2; vang; vang (van gogh)-like 2; Vang1l2; VANGL planar cell polarity protein 2; Vangl2; vangl2 {ECO:0000312; vang-like 2 (van gogh, Drosophila); vang-like protein 2 |
| Gene Symbols | Vangl2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.