Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vangl2 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABN224225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2242-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2242-25UG Supplier MilliporeSigma Supplier No. ABN224225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Vangl2 is a highly specific rabbit polyclonal antibody that targets Vang-like protein 2 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vangl2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols VANGL2,KIAA1215,STB1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Vang-like Protein 2.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_065068
Test Specificity This rabbit polyclonal antibody detects Vang-like protein 2 in human and murine cells. It targets an epitope with in 15 amino acids from the N-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.