Learn More
Vanillic acid, 98%
Description
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.
Solubility
Soluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 210°C to 218°C |
Assay Percent Range | 99% |
Beilstein | 2208364 |
Merck Index | 14,9931 |
Solubility Information | Soluble in water,alcohol and ether. |
Formula Weight | 168.15 |
Percent Purity | 98% |
Quantity | 100 g |
Chemical Name or Material | Vanillic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 204-466-8 |
RTECSNumber | YW5300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.