Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R-like 3/TRPV3 Antibody (N15/39), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212918FR
Description
Vanilloid R-like 3/TRPV3 Monoclonal specifically detects Vanilloid R-like 3/TRPV3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Vanilloid R-like 3/TRPV3 | |
Monoclonal | |
DyLight 680 | |
transient receptor potential cation channel subfamily V member 3, transient receptor potential cation channel, subfamily V, member 3, TrpV3, vanilloid receptor 3, Vanilloid receptor-like 3, vanilloid receptor-related osmotically activated channel protein, VRL3, VRL-3 | |
Mouse | |
Protein G purified | |
RUO | |
162514 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S15-39 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
TRPV3 | |
Synthetic peptide amino acids 458-474 (cytoplasmic C-terminus) of rat TrpV3 | |
0.1 mL | |
Primary | |
Detects approx 70kDa. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction