Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanilloid R1/TRPV1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP197417X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197417X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197417X Supplier Novus Biologicals Supplier No. NBP197417AF488
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative).

Specifications

Antigen Vanilloid R1/TRPV1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220
Gene Symbols TRPV1
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific, Sensory Systems
Primary or Secondary Primary
Gene ID (Entrez) 7442
Target Species Human, Mouse, Rat, Invertebrate, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.