Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197417G
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative).Specifications
Vanilloid R1/TRPV1 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry (Negative) | |
Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Invertebrate, Primate | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRPV1 | |
A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] | |
0.1 mL | |
Neuroscience, Sensory Systems | |
7442 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction