Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171774C
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Vanilloid R1/TRPV1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q8NER1 | |
| TRPV1 | |
| A genomic peptide made to a C-terminal region of the human TRPV1 protein (within residues 725-839). [Swiss-Prot# Q8NER1] | |
| 0.1 mL | |
| Neuroscience, Sensory Systems | |
| 7442 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction