Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanin-1/VNN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP325222
Description
Vanin-1/VNN1 Polyclonal antibody specifically detects Vanin-1/VNN1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Vanin-1/VNN1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
EC 3.5.1, EC 3.5.1.92, HDLCQ8, MGC116930, MGC116931, MGC116932, pantetheinase, Pantetheine hydrolase, Tiff66MGC116933, vanin 1, vanin-1, vannin 1, Vascular non-inflammatory molecule 1 | |
This antibody has been engineered to specifically recognize the recombinant protein Vanin-1/VNN1 using the following amino acid sequence: VFPEVLLSENQLAPGEFQVSTDGRLFSLKPTSGPVLTVTLFGRLYEKDWASNASSGLTAQARIIMLIVIAPIVCSLSW | |
100 μL | |
Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction, Stem Cells | |
8876 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction