Learn More
Invitrogen™ VAP-1 Monoclonal Antibody (TK8-14), eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS1033

Description
Description: VAP-1 (TK8-14) is a mouse monoclonal antibody raised against affinity purifiedVAP-1 from tonsil stroma of human origin. Lymphocyte binding to vascular endothelium is a prerequisite for the move-ment of immune cells from the blood into lymphoid tissues and into sites ofinflammation. Under inflammatory conditions, cell surface expression ofVAP-1 (vascular adhesion protein-1) which is an endothelial sialoglycoprotein,is induced. VAP-1 is a type II transmembrane protein of 84.6 kDa with a singletransmembrane domain and N and O-glycosylation sites in the extracellulardomain. In vivo, VAP-1 exists predominantly as a 180 kDa homodimer andfunctions both as an enzyme (monoamine oxidase) and an adhesion moleculefor lymphocytes. Applications Tested: ELISA, Flow Cytometry, Immunofluorescence, Western Blotting.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Specifications
VAP-1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Q16853 | |
AOC3 | |
Rec. Human VAP-1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
TK8-14 | |
Unconjugated | |
AOC3 | |
amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97 | |
Mouse | |
Affinity chromatography | |
RUO | |
8639 | |
-20°C | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.