Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAP-1 Monoclonal Antibody (TK8-18), eBioscience™
GREENER_CHOICE

Catalog No. 5017701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-177-01 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-177-01 Supplier Invitrogen™ Supplier No. BMS1034
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: Lymphocyte binding to vascular endothelium is a prerequisite for the move-ment of immune cells from the blood into lymphoid tissues and into sites ofinflammation. Under inflammatory conditions, cell surface expression ofVAP-1 (vascular adhesion protein-1) which is an endothelial sialoglycoprotein,is induced. VAP-1 is a type II transmembrane protein of 84.6 kDa with a singletransmembrane domain and N and O-glycosylation sites in the extracellulardomain. In vivo, VAP-1 exists predominantly as a 180 kDa homodimer andfunctions both as an enzyme (monoamine oxidase) and an adhesion moleculefor lymphocytes. Applications Tested: ELISA, Western Blotting.

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAP-1
Applications ELISA, Western Blot
Classification Monoclonal
Clone TK8-18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2-7.4
Gene AOC3
Gene Accession No. Q16853
Gene Alias amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97
Gene Symbols AOC3
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8639
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.