Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-A Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB268282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB268282 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB268282 Supplier Novus Biologicals Supplier No. NBP232249F
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAP-A Polyclonal antibody specifically detects VAP-A in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen VAP-A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias hVAP-33, MGC3745,33 kDa VAMP-associated protein, VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa, VAMP-A, VAP-33, VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD), VAP-AVAMP-associated protein A, vesicle-associated membrane protein-associated protein A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human 33 kDa VAMP-Associated Protein using the numbering given in entry NP_919415.2 (GeneID 9218).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9218
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.