Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAP-B Antibody (103), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289625AF350
Description
VAP-B Monoclonal specifically detects VAP-B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VAP-B | |
Monoclonal | |
Alexa Fluor 350 | |
ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VAP-B (O95292-1; Met1-Pro132). | |
0.1 mL | |
Neuroscience | |
9217 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
103 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction