Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAP-B Antibody (736904), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB58551V
Description
VAP-B Monoclonal antibody specifically detects VAP-B in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOFSpecifications
VAP-B | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
736904 | |
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
E. coli-derived recombinant human VAP-B, Ala2-Pro132, Accession # O95292 | |
0.1 mL | |
Neuroscience | |
9217 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction