Learn More
VAPA Mouse, anti-Human, Unconjugated, Clone: 1F10F9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | VAPA |
---|---|
Clone | 1F10F9 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9892
![]() ![]() |
Proteintech Group Inc
678451IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.
Specifications
VAPA | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat | |
Primary |
1F10F9 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Mouse | |
9218 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.