Learn More
Invitrogen™ VAPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588876
Description
Immunogen sequence: DKLNDMEPSK AVPLNASKQD GPMPKPHSVS LNDTETRKLM EECKRLQGEM MKLSEENRHL RDEGLRLRKV AHSDKPGSTS TASFRDNVTS P; Positive Samples: B-cell, Mouse heart, Mouse pancreas; Cellular Location: Endoplasmic reticulum membrane, Single-pass type IV membrane protein.
The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
VAPA | |
Polyclonal | |
Unconjugated | |
VAPA | |
33 kDa VAMP-associated protein; 33kDa; hVAP-33; synaptobrevin-associated protein of 33kDa; vamp; VAMP (vesicle-associated membrane protein)-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP associated protein A; VAMP-A; VAMP-associated protein A; VAP33; VAP-33; Vapa; VAP-A; vesicle-associated membrane protein, associated protein a; vesicle-associated membrane protein-associated protein A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
30960, 9218 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.41 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9P0L0, Q9WV55 | |
VAPA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 137-227 of human VAPA (NP_9194152). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.