Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552660
Description
Immunogen sequence: NDKLGITPPG NAPTVTSMSS INNTVATPAS YHTKDDPRGL SVLKQEKQKN DMEPSKAVPL NASKQDGPMP KPHSVSLNDT ETRKLMEECK RLQGEMMKLS EENRHLRDEG LRLRKVAHSD KPGSTSTASF Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 84%.
The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
VAPA | |
Polyclonal | |
Unconjugated | |
VAPA | |
33 kDa VAMP-associated protein; 33kDa; hVAP-33; synaptobrevin-associated protein of 33kDa; vamp; VAMP (vesicle-associated membrane protein)-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP associated protein A; VAMP-A; VAMP-associated protein A; VAP33; VAP-33; Vapa; VAP-A; vesicle-associated membrane protein, associated protein a; vesicle-associated membrane protein-associated protein A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0L0 | |
VAPA | |
Recombinant protein corresponding to Human VAPA. Recombinant protein control fragment (Product #RP-90037). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction