Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAPB Mouse anti-Human, Clone: 4F6A6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | VAPB |
---|---|
Clone | 4F6A6 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6625
![]() ![]() |
Proteintech Group Inc
661911IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.Specifications
VAPB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALS8, VAMP associated protein B/C, VAMP B, VAMP B/VAMP C, VAMP C, VAP B, VAP B/VAP C, VAP C, VAPB | |
VAPB | |
IgG1 | |
Protein G | |
Antibody |
4F6A6 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O95292 | |
9217 | |
VAPB Fusion Protein Ag6313 | |
Primary | |
-20°C | |
VAPB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title