Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAPB Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547787
Description
In direct ELISAs, approximately 60% cross-reactivity with recombinant rat VAP-B is observed and less than 5% cross-reactivity with recombinant human VAP-A is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.
Specifications
VAPB | |
Polyclonal | |
Unconjugated | |
VAPB | |
AI225786; AI840687; AI848259; ALS 8; ALS8; ALS8VAMP-B/VAMP-C; D2Abb2e; R75548; UNQ484/PRO983; vamp; VAMP (vesicle-associated membrane protein)-associated protein B and C; Vamp 33b; VAMP associated protein B and C; VAMP B; VAMP B VAMP C; VAMP B/VAMP C; VAMP C; Vamp33b; VAMP-associated 33 kDa protein; VAMP-associated protein 33b; VAMP-associated protein B; VAMPB; VAMP-B; VAMPB/VAMPC; VAMP-B/VAMP-C; VAMPC; VAMP-C; VAP 33b; VAP B; VAP B/VAP C; VAP C; VAP33b; Vapb; VAP-B; VAPB/VAPC; VAP-B/VAP-C; VAPC; VAP-C; vesicle-associated membrane protein, associated protein B and C; vesicle-associated membrane protein-associated protein B; Vesicle-associated membrane protein-associated protein B/C | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
9217 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O95292 | |
VAPB | |
E. coli-derived recombinant human VAP-B Ala2-Pro132. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction