Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAPB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985994
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5994 Supplier Bioss Supplier No. BS4227R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.

Specifications

Antigen VAPB
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VAPB
Gene Accession No. O95292
Gene Alias AI225786; AI840687; AI848259; ALS 8; ALS8; ALS8VAMP-B/VAMP-C; D2Abb2e; R75548; UNQ484/PRO983; VAMP (vesicle-associated membrane protein)-associated protein B and C; Vamp 33b; VAMP associated protein B and C; VAMP B; VAMP B VAMP C; VAMP B/VAMP C; VAMP C; Vamp33b; VAMP-associated 33 kDa protein; VAMP-associated protein 33b; VAMP-associated protein B; VAMPB; VAMP-B; VAMPB/VAMPC; VAMP-B/VAMP-C; VAMPC; VAMP-C; VAP 33b; VAP B; VAP B/VAP C; VAP C; VAP33b; Vapb; VAP-B; VAPB/VAPC; VAP-B/VAP-C; VAPC; VAP-C; vesicle-associated membrane protein, associated protein B and C; Vesicle-associated membrane protein-associated protein B; Vesicle-associated membrane protein-associated protein B/C
Gene Symbols VAPB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VAPB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.