Learn More
Invitrogen™ VAPB Recombinant Rabbit Monoclonal Antibody (103)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529638
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human VAPB/VAP-B.
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.
Specifications
VAPB | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O95292 | |
VAPB | |
Recombinant Human VAPB/VAP-B Protein (Met1-Pro132). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
103 | |
Unconjugated | |
VAPB | |
AI225786; AI840687; AI848259; ALS 8; ALS8; ALS8VAMP-B/VAMP-C; D2Abb2e; R75548; UNQ484/PRO983; vamp; VAMP (vesicle-associated membrane protein)-associated protein B and C; Vamp 33b; VAMP associated protein B and C; VAMP B; VAMP B VAMP C; VAMP B/VAMP C; VAMP C; Vamp33b; VAMP-associated 33 kDa protein; VAMP-associated protein 33b; VAMP-associated protein B; VAMPB; VAMP-B; VAMPB/VAMPC; VAMP-B/VAMP-C; VAMPC; VAMP-C; VAP 33b; VAP B; VAP B/VAP C; VAP C; VAP33b; Vapb; VAP-B; VAPB/VAPC; VAP-B/VAP-C; VAPC; VAP-C; vesicle-associated membrane protein, associated protein B and C; vesicle-associated membrane protein-associated protein B; Vesicle-associated membrane protein-associated protein B/C | |
Rabbit | |
Protein A | |
RUO | |
9217 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.