Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Flexible Tubing
VapLock™ Adapters for Flexible Tubing are designed for secure leak-resistant connections
$18.58 - $22.24
Specifications
| Color | Gray |
|---|---|
| For Use With (Application) | Solvent Safety |
| For Use With (Equipment) | VapLock Solvent Containment Systems |
| Material | Polypropylene |
| Catalog Number | Mfr. No. | Size | Thread Size (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Thread Size (English) | Price | Quantity & Availability | ||||
|
13-311-220
|
Cole-Parmer™
1202018 |
1/8 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $21.30
|
|
||||
|
13-311-222
|
Cole-Parmer™
1202020 |
1/4 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $19.41
|
|
||||
|
13-311-223
|
Cole-Parmer™
1202021 |
5/16 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $20.30
|
|
||||
|
13-311-224
|
Cole-Parmer™
1202022 |
3/8 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $19.90
|
|
||||
|
13-311-225
|
Cole-Parmer™
1202023 |
1/2 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $19.07
|
|
||||
|
13-311-226
|
Cole-Parmer™
1202024 |
5/32 in. Barb x NPT(M) | 1/4 in. NPT(M) |
Pack of 10 for $18.86
|
|
||||
|
13-311-228
|
Cole-Parmer™
1202026 |
1/8 in. Barb x NPT(M) | 1/8 in. NPT(M) |
Pack of 10 for $20.74
|
|
||||
|
13-311-230
|
Cole-Parmer™
1202028 |
1/4 in. Barb x NPT(M) | 1/8 in. NPT(M) |
Pack of 10 for $19.66
|
|
||||
|
13-311-231
|
Cole-Parmer™
1202029 |
5/16 in. Barb x NPT(M) | 1/8 in. NPT(M) |
Pack of 10 for $18.86
|
|
||||
|
13-311-232
|
Cole-Parmer™
1202030 |
3/8 in. Barb x NPT(M) | 1/8 in. NPT(M) |
Pack of 10 for $18.58
|
|
||||
|
13-311-233
|
Cole-Parmer™
1202031 |
1/4 in. Barb x NPT(M) | 1/2 in. NPT(M) |
Pack of 10 for $20.84
|
|
||||
|
13-311-234
|
Cole-Parmer™
1202032 |
3/8 in. Barb x NPT(M) | 1/2 in. NPT(M) |
Pack of 10 for $20.30
|
|
||||
|
13-311-236
|
Cole-Parmer™
1202034 |
5/8 in. Barb x NPT(M) | 1/2 in. NPT(M) |
Pack of 10 for $22.24
|
|
||||
|
13-311-237
|
Cole-Parmer™
1202035 |
3/4 in. Barb x NPT(M) | 1/2 in. NPT(M) |
Pack of 10 for $20.30
|
|
||||
|
13-311-221
|
Cole-Parmer™
1202019 |
3/16 in. Barb x NPT(M) | 1/4 in. NPT(M) | N/A | N/A | N/A | |||
|
13-311-227
|
Cole-Parmer™
1202025 |
1/16 in. Barb x NPT(M) | 1/8 in. NPT(M) | N/A | N/A | N/A | |||
Description
Use with flexible, soft-wall tubingWarranty and Services
None
Specifications
| Gray | |
| Solvent Safety | |
| VapLock Solvent Containment Systems | |
| Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title