Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Adapters with Large Bore for Semi-Rigid Tubing

VapLock™ Adapters with Large Bore for Semi-Rigid Tubing are designed for secure leak-resistant connections

$33.16 - $42.97

Specifications

For Use With (Application) Solvent Safety
For Use With (Equipment) VapLock Solvent Containment Systems
Material Polypropylene
View More Specs

Products 5
Catalog Number Mfr. No. Size Thread Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Thread Size (English) Price Quantity & Availability  
13-311-239
View Documents
Cole-Parmer™
1202037
5/16 in. OD Compression x NPT(M) 1/4 in. NPT(M)
Pack of 5 for $35.79
Only null left
Add to Cart
 
13-311-240
View Documents
Cole-Parmer™
1202038
3/8 in. OD Compression x NPT(M) 1/4 in. NPT(M)
Pack of 5 for $35.79
Only null left
Add to Cart
 
13-311-241
View Documents
Cole-Parmer™
1202039
1/2 in. OD Compression x NPT(M) 1/4 in. NPT(M)
Pack of 5 for $42.97
Only null left
Add to Cart
 
13-311-242
View Documents
Cole-Parmer™
1202040
3/8 in. OD Compression x NPT(M) 1/2 in. NPT(M)
Pack of 5 for $33.29
Only null left
Add to Cart
 
13-311-243
View Documents
Cole-Parmer™
1202041
1/2 in. OD Compression x NPT(M) 1/2 in. NPT(M)
Pack of 5 for $33.16
Only null left
Add to Cart
 
Description

Description

Integral ferrule prevents loss of small parts

Warranty and Services

None

Specifications

Specifications

Solvent Safety
VapLock Solvent Containment Systems
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.