Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Reversible Valves for Solvent Management

VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling

$85.14 - $121.70

Specifications

Type Reversible Valve
For Use With (Application) Solvent Safety
For Use With (Equipment) VapLock Solvent Containment Systems
Pressure 0.1 to 0.25 psi crack pressure
Thread Size (English) 2 x 1/4 in. NPT(M) x 1/4 in. NPT(M)
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
13-311-262
View Documents
Cole-Parmer™
1202060
Polypropylene, FFKM
Each for $121.70
Only null left
Add to Cart
 
13-311-260
View Documents
Cole-Parmer™
1202058
Polypropylene, EPDM
Each for $90.65
Only null left
Add to Cart
 
13-311-261
View Documents
Cole-Parmer™
1202059
Polypropylene, Viton
Each for $85.14
Only null left
Add to Cart
 
Description

Description

  • Minimize evaporation and relieve pressure to improve safety
  • Change orientation to switch between inlet and outlet flow regulation

Warranty and Services

None

Specifications

Specifications

Reversible Valve
VapLock Solvent Containment Systems
2 x 1/4 in. NPT(M) x 1/4 in. NPT(M)
Solvent Safety
0.1 to 0.25 psi crack pressure
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.