Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex™ VapLock™ Solvent Delivery Cap Kits

Reduce chemical evaporation and exposure to hazardous vapors on multiple carboys

$381.42 - $423.73

Specifications

Includes Four color-coded GL45 safety caps in blue, red, yellow, and green, with eight connections for 1/8 in. OD tubing, one connection for 2.2 mm OD tubing, and four PTFE plugs.
Thread Size GL45
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
12-020-91
View Documents
Spex™
1202091
Solvent Delivery Cap Kit, Reverse Phase
Each for $381.42
Only null left
Add to Cart
 
12-020-92
View Documents
Spex™
1202092
Solvent Delivery Cap Kit, Normal Phase
Each for $423.73
Only null left
Add to Cart
 
Description

Description

  • Less expensive than ordering individual components
  • Color coded caps allow easy solvent identification
  • Choose from FFKM, EPDM, or Viton™ valve seal options
  • Ideal for quaternary systems
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4''-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable. Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 μm PTFE filter to reduce particulate contamination.
Specifications

Specifications

Four color-coded GL45 safety caps in blue, red, yellow, and green, with eight connections for 1/8 in. OD tubing, one connection for 2.2 mm OD tubing, and four PTFE plugs.
GL45
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.