Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
$72.16 - $139.26
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | Solvent-Compatible Containers |
Thread Size (English) | 1/4 in.-28 UNF(F) |
Catalog Number | Mfr. No. | Size | Material | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Material | Type | Price | Quantity & Availability | |||
13-311-006
|
Cole-Parmer™
1201806 |
GL45 | Green/white PTFE, PP, SS, EPDM | Two 1/4 in.-28 ports |
Each for $90.70
|
|
|||
13-311-007
|
Cole-Parmer™
1201807 |
GL45 | Green/white PTFE, PP, SS, EPDM | Four 1/4 in.-28 ports |
Each for $88.57
|
|
|||
13-311-008
|
Cole-Parmer™
1201808 |
GL45 | Yellow/white PTFE, PP, SS, FFKM | Two 1/4 in.-28 ports |
Each for $130.52
|
|
|||
13-311-009
|
Cole-Parmer™
1201809 |
GL45 | Yellow/white PTFE, PP, SS, FFKM | Four 1/4 in.-28 ports |
Each for $139.26
|
|
|||
13-311-010
|
Cole-Parmer™
1201810 |
GL45 | Blue/white PTFE, PP, SS, Viton | Two 1/4 in.-28 ports |
Each for $77.96
|
|
|||
13-311-011
|
Cole-Parmer™
1201811 |
GL45 | Blue/white PTFE, PP, SS, Viton | Four 1/4 in.-28 ports |
Each for $102.92
|
|
|||
13-311-018
|
Cole-Parmer™
1201818 |
GL38 | Black/white PTFE, PP, SS, FFKM | Two 1/4 in.-28 ports |
Each for $107.29
|
|
|||
13-311-019
|
Cole-Parmer™
1201819 |
GL38 | Black/white PTFE, PP, SS, EPDM | Two 1/4 in.-28 ports |
Each for $72.16
|
|
|||
13-311-020
|
Cole-Parmer™
1201820 |
GL38 | Black/white PTFE, PP, SS, Viton | Two 1/4 in.-28 ports |
Each for $72.16
|
|
|||
13-311-021
|
Cole-Parmer™
1201821 |
GL38 | Black/white PTFE, PP, SS, FFKM | Four 1/4 in.-28 ports |
Each for $133.73
|
|
|||
13-311-022
|
Cole-Parmer™
1201822 |
GL38 | Black/white PTFE, PP, SS, EPDM | Four 1/4 in.-28 ports |
Each for $84.14
|
|
|||
13-311-023
|
Cole-Parmer™
1201823 |
GL38 | Black/white PTFE, PP, SS, Viton | Four 1/4 in.-28 ports |
Each for $86.28
|
|
|||
13-311-030
|
Cole-Parmer™
1201830 |
Nalgene 38-430 | White PTFE, PP, SS, EPDM | Two 1/4 in.-28 ports |
Each for $74.38
|
|
|||
13-311-031
|
Cole-Parmer™
1201831 |
Nalgene 38-430 | White PTFE, PP, SS, EPDM | Four 1/4 in.-28 ports |
Each for $78.23
|
|
|||
13-311-032
|
Cole-Parmer™
1201832 |
Nalgene 38-430 | White PTFE, PP, SS, FFKM | Two 1/4 in.-28 ports |
Each for $101.76
|
|
|||
13-311-033
|
Cole-Parmer™
1201833 |
Nalgene 38-430 | White PTFE, PP, SS, FFKM | Four 1/4 in.-28 ports |
Each for $115.03
|
|
|||
13-311-034
|
Cole-Parmer™
1201834 |
Nalgene 38-430 | White PTFE, PP, SS, Viton | Two 1/4 in.-28 ports |
Each for $82.70
|
|
|||
13-311-035
|
Cole-Parmer™
1201835 |
Nalgene 38-430 | White PTFE, PP, SS, Viton | Four 1/4 in.-28 ports |
Each for $81.09
|
|
|||
13-311-042
|
Cole-Parmer™
1201842 |
Wako 38 mm | White PTFE, PP, SS, EPDM | Two 1/4 in.-28 ports |
Each for $79.14
|
|
|||
13-311-044
|
Cole-Parmer™
1201844 |
Wako 38 mm | White PTFE, PP, SS, FFKM | Two 1/4 in.-28 ports |
Each for $100.67
|
|
|||
13-311-045
|
Cole-Parmer™
1201845 |
Wako 38 mm | White PTFE, PP, SS, FFKM | Four 1/4 in.-28 ports |
Each for $112.86
|
|
|||
13-311-046
|
Cole-Parmer™
1201846 |
Wako 38 mm | White PTFE, PP, SS, Viton | Two 1/4 in.-28 ports |
Each for $88.46
|
|
|||
13-311-047
|
Cole-Parmer™
1201847 |
Wako 38 mm | White PTFE, PP, SS, Viton | Four 1/4 in.-28 ports |
Each for $84.09
|
|
|||
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODWarranty and Services
None
Specifications
Solvent Safety | |
Solvent-Compatible Containers | |
1/4 in.-28 UNF(F) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title