Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Solvent Waste Kits for Drums and Panel Mounting

VapLock™ Solvent Waste Kits for Drums and Panel Mounting collect effluent from multiple sources while minimizing harmful solvent exhaust

$542.62 - $902.11

Specifications

Type Solvent Waste Kit
For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
Thread Size (English) 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
View More Specs

Products 4
Catalog Number Mfr. No. Size Material Price Quantity & Availability  
Catalog Number Mfr. No. Size Material Price Quantity & Availability  
13-311-167
View Documents
Cole-Parmer™
1201965
2 in. Fine Thread Polypropylene
Each for $578.55
Only null left
Add to Cart
 
13-311-173
View Documents
Cole-Parmer™
1201971
2 in. Coarse Thread Polypropylene
Each for $543.78
Only null left
Add to Cart
 
13-311-179
View Documents
Cole-Parmer™
1201977
Panel Mount Polypropylene
Each for $542.62
Only null left
Add to Cart
 
13-311-168
View Documents
Cole-Parmer™
1201966
2 in. Fine Thread PTFE
Each for $902.11
Only null left
Add to Cart
 
Description

Description

Various cap sizes available to fit your solvent container

Warranty and Services

None

Specifications

Specifications

Solvent Waste Kit
Solvent Safety
Solvent-Compatible Containers
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.