Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit and Six Ports 1/8 in.
Shop All Justrite Safety Group Products

Click to view available options
Capacity (Metric):
13.5 L
20 L
Description
- For packaging group II and hazardous chemicals
- Carbon filter prevents volatile organic compound vapors
- Two-piece cap design
Specifications
Specifications
| Capacity (English) | 3.6 gal. |
| Capacity (Metric) | 13.5 L |
| Certifications/Compliance | UN/DOT |
| Closure Material | Polypropylene |
| Closure Size | 70 mm |
| Color | White |
| Description | Six (6) 1/8 in. Tubing OD Ports |
| Dimensions (L x W x H) | 287 x 235 x 333 mm |
| For Use With (Application) | Safe transport of hazardous liquids |
| Height (English) | 13.1 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction