Learn More
Justrite™ VaporTrap™ Polypropylene, UN/DOT-Certified Cap with Filter Kit, Four Ports 1/8 in. OD Tubing, Three Ports 1/4 in. OD Tubing
GREENER_CHOICE
Catalog No. 19830405 Shop All Justrite Safety Group Products
Change view
Click to view available options
Description:
Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Catalog No. Description
19-830-405 Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
1 options
Catalog No. 19-830-405 Supplier Justrite™ Supplier No. 12833
Add to Cart
Add to Cart
For Use With: carboys with 70mm cap
  • For packaging group II and hazardous chemicals
  • Carbon filter prevents volatile organic compound vapors
Specifications
Description Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Dimensions (L x W x H) 3.5 x 9.5 x 3.75 in.
For Use With (Equipment) carboys with 70mm cap
Material Polypropylene
Product Type Cap