Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Varenicline tartrate
GSA_VAGREENER_CHOICE

Catalog No. 375410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-541-0 10 mg
37-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-541-0 Supplier Tocris Bioscience™ Supplier No. 3754/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective α4β2 partial agonist; orally active

Orally active, subtype-selective partial agonist at α4β2 nicotinic receptors (Ki values are 0.06, 240, 322 and 3540 nM for α4β2, α3β4, α7, α1βγδ receptors respectively). Reduces nicotine-evoked dopamine release in vitro and decreases nicotine self-administration in vivo. Also potently activates PSAM4-5-HT3 and PSAM4-GlyR chimeric ion channels (EC50 values are 4 and 1.6 nM, respectively)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 375815-87-5
Molecular Formula C17H19N3O6

Specifications

Quantity 10 mg
Formula Weight 361.37
Purity >99%
Molecular Weight (g/mol) 361.354
Chemical Name or Material Varenicline tartrate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.