Learn More
Variable Inertia Kit
Click to view available options
:
Variable Inertia Kit
Description
Includes
eight steel balls (3/4 in. dia.), two plastic discs with hardware (3 1/2 in. dia.), and instructions.
Easily change the distribution of mass by slipping solid steel balls into various disc compartments. Observe what happens when the differently weighted discs roll down an incline.
Specifications
Specifications
| Product Type | Kit, Variable Inertia |
| Includes | Eight Steel balls, two Plastic discs with hardware and instructions |