Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Variable Micropipets
SDPSCI_ED

Catalog No. p-3032137 Shop All EDVOTEK Products
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 5000 μL
2 to 20 μL
20 to 200 μL
5 to 50 μL

Catalog No. S90243B

Supplier: Edvotek™ 589

Only null left
Add to Cart
Add to Cart

Includes

  • Service wrench
  • Instructions

Automatic tip ejector makes operation fast and convenient

EDVOTEK™'s new line of Variable Micropipets are smartly designed with volumes spanning 0.1 to 5000 μl. The contoured grip, lightweight and low force design, and large volume display make them easy-to-use. Sample volumes are selected by twisting the top and sample delivery utilizes standard micropipet tips. Each micropipet is fully autoclavable and calibratable. Make your laboratory experience authentic with these durable micropipets backed by a lifetime warranty.

Specifications

Autoclavable Autoclavable
Color Gray
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Plastic
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Variable Micropipet
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
Includes Service wrench, Instructions
Warranty Lifetime
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.