Learn More
Accumax Lab Technology Variable Micropipets
SCI_ED
Catalog No. S902432K
Click to view available options
Volume (Metric):
0.5 to 10 μL
1000 to 5000 μL
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Service wrench and instructions
Automatic tip ejector makes operation fast and convenient.
  • Ergonomically designed
  • Digital display indicates the chosen volume
  • Deliver precise and reproducible volumes with ease
  • Utilize standard disposable micropipet tips (Except the 1000-5000μL model, S902432K)
  • No calibration servicing needed
Warranty and Services
30 days
Specifications
Accuracy Inaccuracy of ±0.8 μL for 1000 μL, or ±0.5% for 5000 μL
Type Variable Micropipet
Volume (Metric) 1000 to 5000 μL
Finger Style Ergonomic
Includes Service wrench, instructions
Increments 50μL
Warranty 30 Days
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.