Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113815 Supplier Invitrogen™ Supplier No. PA5113815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQRLRERRAA SGYPVKVPLE VQEADEAKLQ QTEAELRKVD EAIALFQKML For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. This protein belongs to the class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VARS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VARS1
Gene Accession No. P26640
Gene Alias Bat6; D17H6S56E; EC 6.1.1.9; G7A; Protein G7a; TrsVal; valine tRNA ligase 1, cytoplasmic; valine--tRNA ligase; ValRS; valyl trna synthetase; valyl-tRNA synthetase; valyl-tRNA synthetase 2; VARS; VARS1; VARS2
Gene Symbols VARS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human VARS.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7407
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.