Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VARS Polyclonal Antibody

Supplier: Thermo Scientific PA5118347
This item is not returnable.
View return policy
Description
Specifications
| VARS | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P26640 | |
| VARS1 | |
| E. coli-derived Human VARS fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| VARS1 | |
| Bat6; D17H6S56E; EC 6.1.1.9; G7A; Protein G7a; TrsVal; valine tRNA ligase 1, cytoplasmic; valine--tRNA ligase; ValRS; valyl trna synthetase; valyl-tRNA synthetase; valyl-tRNA synthetase 2; VARS; VARS1; VARS2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 7407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction