Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VARS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566493
Description
Immunogen sequence: NALRFILNALG EKFVPQPAEE LSPSSPMDAW ILSRLALAAQ ECERGFLTRE LSLVTHALHH FWLHNLCDVY LE Highest antigen sequence identity to the following orthologs - mouse 85%, rat 85%.
This gene encodes a mitochondrial aminoacyl-tRNA synthetase, which catalyzes the attachment of valine to tRNA(Val) for mitochondrial translation. Mutations in this gene cause combined oxidative phosphorylation deficiency-20, and are also associated with early-onset mitochondrial encephalopathies. Alternative splicing of this gene results in multiple transcript variants.
Specifications
VARS2 | |
Polyclonal | |
Unconjugated | |
VARS2 | |
1190004I24Rik; COXPD20; Kiaa1885; mKIAA1885; valine tRNA ligase 2, mitochondrial (putative); valine--tRNA ligase, mitochondrial; ValRS; valyl trna synthetase; Valyl-tRNA synthetase; valyl-tRNA synthetase 2, mitochondrial; valyl-tRNA synthetase 2, mitochondrial (putative); valyl-tRNA synthetase like; valyl-tRNA synthetase, mitochondrial; Valyl-tRNA synthetase-like; VARS2; VARS2L; VARSL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5ST30 | |
VARS2 | |
Recombinant Human VARS2. Recombinant protein control fragment (Product #RP-103920). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction