Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vars2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0916 Supplier Bioss Supplier No. BS10504R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a mitochondrial aminoacyl-tRNA synthetase, which catalyzes the attachment of valine to tRNA(Val) for mitochondrial translation. Mutations in this gene cause combined oxidative phosphorylation deficiency-20, and are also associated with early-onset mitochondrial encephalopathies. Alternative splicing of this gene results in multiple transcript variants.

Specifications

Antigen vars2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VARS2
Gene Accession No. Q5ST30
Gene Alias 1190004I24Rik; COXPD20; Kiaa1885; mKIAA1885; valine tRNA ligase 2, mitochondrial (putative); Valine--tRNA ligase, mitochondrial; ValRS; Valyl-tRNA synthetase; valyl-tRNA synthetase 2, mitochondrial; valyl-tRNA synthetase 2, mitochondrial (putative); valyl-tRNA synthetase like; valyl-tRNA synthetase, mitochondrial; Valyl-tRNA synthetase-like; VARS2; VARS2L; VARSL
Gene Symbols VARS2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human vars2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57176
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.