Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VASA Polyclonal Antibody, Abbomax

Catalog No. 502203969
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3969 Supplier Abbomax Inc Supplier No. 601350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse embryonic germ line cell Cellular location: Cytoplasmic.

VASA Polyclonal antibody specifically detects VASA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen VASA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene DDX4
Gene Accession No. Q9NQI0
Gene Alias ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the epitope GDGVGG-S-GGEGGGY WITHOUT the phosphorylated serine of human VASA protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54514
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.