Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vascular Endothelium Marker Antibody (10), Novus Biologicals™

Mouse Monoclonal Antibody
$173.10
Specifications
Antigen | Vascular Endothelium Marker |
---|---|
Clone | 10 |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:10 - 1:500 |
Applications | Immunohistochemistry |
Description
Vascular Endothelium Marker Monoclonal antibody specifically detects Vascular Endothelium Marker in Human samples. It is validated for ImmunohistochemistrySpecifications
Vascular Endothelium Marker | |
1 mg/mL | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer, Cardiovascular Biology | |
PBS | |
IgM | |
Thiophilic Chromatography |
10 | |
Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Fibronectin-purified human monocytes. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title