Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Vasculin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540615
Description
Peptide sequence: RKEKNGWRTH GRNGTENINH RGGYHGGSSR SRSSIFHAGK SQGLHENNIP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.
Vasculin is a novel vascular protein differentially expressed in human atherogenesis.
Specifications
Vasculin | |
Polyclonal | |
Unconjugated | |
GPBP1 | |
1700034P14Rik; AU019836; D230035M11Rik; DKFZP761C169; GC-rich promoter binding protein 1; GC-rich promoter-binding protein 1; GPBP; GPBP1; LOW QUALITY PROTEIN: vasculin; mGPBP; RGD1305492; SSH6; Unknown (protein for MGC:139708); vascular wall-linked protein; VASCULIN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
65056 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WP2 | |
GPBP1 | |
Synthetic peptide directed towards the N-terminal of human DKFZP761C169 (aa 67-116). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction