Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VASH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596518 Supplier Invitrogen™ Supplier No. PA596518
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPGGKKVAGG GSSGATPTSA AATAPSGVRR LETSEGTSAQ RDEEPEEEGE EDLRDGGVPF FVNRGGLPVD EATWERMWKH VAKIHPDGEK VAQRIRGATD LPKIPIPSVP TFQPSTPVPE RLEAVQRYIR ELQYNHTGTQ FFEIKKSRPL TGLMDLAKEM TKEALPIKCL EAVILGIYLT NSMPTLERFP ISFKTYFSGN YFRHIVLGVN FAGRYGALGM SRREDLMYKP PAFRTLSELV LDFEAAYGRC WHVLKKVKLG QSVSHDPHSV EQIEWKHSVL DVERLGRDDF RKELERHARD MRLKIGKGTG PPSPTKDRKK DVSSPQRAQS SPHRRNSRSE RRPSGDKKTS EPKAMPDLNG YQIRV; Positive Samples: THP-1, Mouse brain, Mouse stomach.

VASH1 is a protein that possesses actin binding activity and metallocarboxypeptidase activity. It is involved in several processes including negative regulation of angiogenesis, negative regulation of blood vessel endothelial cell migration, and proteolysis. VASH1 acts upstream of or within processes such as negative regulation of endothelial cell migration, negative regulation of endothelial cell proliferation, and negative regulation of lymphangiogenesis. It is located in the apical part of the cell, endoplasmic reticulum, and extracellular space. VASH1 is implicated in liver cirrhosis and portal hypertension, and is a biomarker of liver cirrhosis. Diseases associated with VASH1 include Portal Hypertension and Myasthenic Syndrome, Congenital, 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VASH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VASH1
Gene Accession No. Q7L8A9, Q8C1W1
Gene Alias AI834978; D930046M13Rik; G630009D10Rik; KIAA1036; RGD1564082; TTCP 1; Tubulin carboxypeptidase 1; Tubulinyl-Tyr carboxypeptidase 1; Tyrosine carboxypeptidase 1; VASH; VASH1; vasohibin 1; vasohibin-1
Gene Symbols VASH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-365 of human VASH1 (NP_0557241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22846, 238328
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.