Learn More
VASH1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | VASH1 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8208
![]() ![]() |
Proteintech Group Inc
127301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
12730-1-AP detects 41 kDa form secreted protein and 36 kDa form protein accumulated within the cells or pericellular milieu in SDS-PAGE.
VASH1 was originally identified as an endothelium-derived vascular endothelial growth factor (VEGF)-inducible angiogenesis inhibitor that acts in a negative feedback manner. VASH1 mRNA is expressed brain, heart, kidney and placenta in the adult, with robust expression in various organs during embryonic development. VASH1 expression in tumor cells and tumor vasculature is silenced by methylation by increased expression of Zeste homolog 2 (EZH2). The increase of EZH2 expression is induced by VEGF stimulation, leading to poor clinical prognosis. Recombit adenovirus expressing VASH prevented tumor angiogenesis and inhibited tumor growth, suggesting that it may be a potentially valuable antitumor therapy in the clinic.Specifications
VASH1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q7L8A9 | |
22846 | |
VASH1 Fusion Protein Ag3435 | |
Primary | |
-20°C | |
VASH1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA1036, VASH, VASH1, vasohibin 1 | |
VASH1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.