Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VASH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571863
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571863 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571863 Supplier Invitrogen™ Supplier No. PA571863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VASH2 Polyclonal Antibody for Western Blot, Flow

VASH2 is a angiogenesis inhibitor. It inhibits network formation by endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VASH2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Vash2
Gene Accession No. Q86V25, Q8C5G2
Gene Alias B130052G07Rik; RGD1564105; testicular tissue protein Li 222; Tubulinyl-Tyr carboxypeptidase 2; Vash2; Vashl; vasohibin 2; vasohibin-2; Vasohibin-like protein
Gene Symbols Vash2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 216-245 amino acids from the C-terminal region of human VASH2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226841, 79805
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.