Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vasohibin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220845
Description
Vasohibin Polyclonal specifically detects Vasohibin in Human samples. It is validated for Western Blot.Specifications
Vasohibin | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
KIAA1036vasohibin-1, VASH, vasohibin 1 | |
Rabbit | |
41 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q7L8A9 | |
VASH1 | |
Recombinant protein encompassing a sequence within the center region of human VASH1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
22846 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction