Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vasorin/SLIT-like 2 Antibody (010), Alexa Fluor™ 750 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290245AF750
Description
Vasorin/SLIT-like 2 Monoclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for ELISA, ELISASpecifications
Vasorin/SLIT-like 2 | |
Monoclonal | |
Alexa Fluor 750 | |
Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Vasorin/SLIT-like 2 (Q6EMK4; Met1-Pro575). | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers | |
114990 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
010 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction