Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vasorin/SLIT-like 2 Antibody (010), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290245F
Description
Vasorin/SLIT-like 2 Monoclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for ELISA, ELISASpecifications
| Vasorin/SLIT-like 2 | |
| Monoclonal | |
| FITC | |
| Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Vasorin/SLIT-like 2 (Q6EMK4; Met1-Pro575). | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 114990 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 010 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction