Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vasorin/SLIT-like 2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318471C
Description
Vasorin/SLIT-like 2 Polyclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western BlotSpecifications
Vasorin/SLIT-like 2 | |
Polyclonal | |
Western Blot | |
Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin | |
Recognizes region between residues Between between 623 and 673.. | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers | |
114990 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction