Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VASP Antibody (OTI4D6), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274830PECY7
Description
VASP Monoclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
VASP | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
VASP | |
Full length recombinant protein of human VASP (NP_003361) produced in 293T Cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI4D6 | |
Flow Cytometry | |
vasodilator-stimulated phosphoprotein | |
Mouse | |
Affinity Purified | |
Phospho Specific | |
7408.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction