Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VASP, Mouse anti-Human, Clone: IE273, Abnova™

Catalog No. 89998864 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-864 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-864 Supplier Abnova Corporation Supplier No. MAB16120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen VASP
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone IE273
Conjugate Unconjugated
Dilution Immunocytochemistry (1:100-500) Western Blot (1:2000-50000) The optimal working dilution should be determined by the end user.
Formulation In PBS (2 mg/mL BSA, 0.02% sodium azide).
Gene vasodilator-stimulated phosphoprotein
Gene Alias -
Gene Symbols VASP
Host Species Mouse
Immunogen Purified human VASP.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7408
Test Specificity Recognizes both the Ser157 dephosphorylated and Ser157 phosphorylated form of VASP.
Target Species Bovine, Human, Pig, Rabbit, Sheep
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.