Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VASP, p Thr278 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323264
Description
VASP Polyclonal antibody specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
VASP | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
vasodilator-stimulated phosphoprotein | |
Phospho-VASP (Thr-278) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding threonine 278 in human VASP. This sequence has significant homology to the conserved site in rat and mouse VASP, but is not conserved in mena or EVL proteins. [UniProt# P50552] | |
0.1 mL | |
Phospho Specific | |
7408 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction