Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543777 Supplier Invitrogen™ Supplier No. PA543777
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PPAPGPGQLT LRLRACGLNF ADLMARQGLY DRLPPLPVTP GMEGAGVVIA Sequence homology: Cow: 93%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%; Zebrafish: 86%.

Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VAT1
Gene Accession No. Q99536
Gene Alias FLJ20230; membrane protein of cholinergic synaptic vesicles; mitofusin-binding protein; Protein MIB; si:dz163l24.2; solute carrier family 18 (vesicular monoamine), member 1; synaptic vesicle membrane protein VAT-1 homolog; VAT1; Vat-1; vat1 protein; VATI; vesicle amine transport 1; vesicle amine transport protein 1 homolog (T californica); vesicle amine transport protein 1 homolog (T. californica)
Gene Symbols VAT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human VAT1 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10493
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.